siRNA Details
| Gene Symbol: THAP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317776.1 | NP_001304705.1 | 5 | 818 | ||
| NM_024672.5 | NP_078948.3 | 4 | 752 | |||
| XM_005263220.2 | XP_005263277.1 | 4 | 709 | |||
| XM_011532246.2 | XP_011530548.1 | 3 | 427 | |||
| XM_017008603.1 | XP_016864092.1 | 4 | 1592 | |||
| XM_017008604.1 | XP_016864093.1 | 4 | 1645 | |||
| XM_017008605.1 | XP_016864094.1 | 4 | 686 | |||
| XM_017008606.1 | XP_016864095.1 | 5 | 810 | |||
| XM_017008607.1 | XP_016864096.1 | 4 | 1025 | |||
| XM_017008608.1 | XP_016864097.1 | 5 | 1664 | |||
| XM_017008609.1 | XP_016864098.1 | 4 | 699 | |||
| XM_017008610.1 | XP_016864099.1 | 4 | 686 | |||
| GenBank mRNA | AK026973.1 | - | 2 | 82 | ||
| AK091412.1 | - | 4 | 649 | |||
| AK225333.1 | - | 2 | 89 | |||
| BC131722.1 | - | 5 | 699 | |||
| BX648134.1 | - | 4 | 857 | |||
| DB154392.1 | - | 2 | 227 | |||

THAP9
Genomic Map