siRNA Details
| Gene Symbol: SH3D21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162530.1 | NP_001156002.1 | 14 | 1575 | ||
| NM_024676.4 | NP_078952.4 | 11 | 1381 | |||
| XM_011542153.2 | XP_011540455.1 | 14 | 1620 | |||
| XM_017002340.1 | XP_016857829.1 | 14 | 1620 | |||
| XM_017002341.1 | XP_016857830.1 | 14 | 1620 | |||
| XM_017002342.1 | XP_016857831.1 | 14 | 1620 | |||
| XM_017002343.1 | XP_016857832.1 | 14 | 1620 | |||
| XM_017002344.1 | XP_016857833.1 | 14 | 1620 | |||
| XM_017002345.1 | XP_016857834.1 | 14 | 1620 | |||
| XM_017002346.1 | XP_016857835.1 | 6 | 992 | |||
| GenBank mRNA | AK026591.1 | - | 1 | 44 | ||
| AK056459.1 | - | 7 | 1076 | |||
| AK297015.1 | - | 11 | 1378 | |||
| BC048273.1 | - | 1 | 81 | |||
| BC101676.1 | - | 3 | 622 | |||
| BC101678.1 | - | 2 | 569 | |||

SH3D21
Genomic Map