siRNA Details
| Gene Symbol: KCTD17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282684.1 | NP_001269613.1 | 3 | 377 | ||
| NM_001282685.1 | NP_001269614.1 | 3 | 377 | |||
| NM_001282686.1 | NP_001269615.1 | 3 | 377 | |||
| NM_024681.3 | NP_078957.2 | 3 | 377 | |||
| XM_005261741.3 | XP_005261798.2 | 3 | 388 | |||
| XM_005261742.2 | XP_005261799.2 | 3 | 388 | |||
| XM_005261743.2 | XP_005261800.2 | 3 | 388 | |||
| XM_005261744.2 | XP_005261801.1 | 3 | 388 | |||
| XM_011530374.2 | XP_011528676.1 | 3 | 388 | |||
| XM_011530377.2 | XP_011528679.1 | 3 | 388 | |||
| GenBank mRNA | AK022304.1 | - | 3 | 353 | ||
| AK308720.1 | - | 3 | 626 | |||
| BC025403.1 | - | 3 | 376 | 4824153 | ||
| BC031038.1 | - | 3 | 360 | 5277700 | ||
| BG832241.1 | - | 3 | 358 | |||
| BP362002.1 | - | 3 | 374 | |||
| BX463485.2 | - | 3 | 237 | |||
| BX951630.1 | - | 3 | 289 | |||
| DA258908.1 | - | 3 | 370 | |||
| HY029628.1 | - | 4 | 302 | |||
| HY034580.1 | - | 3 | 376 | |||

KCTD17
Genomic Map