siRNA Details
| Gene Symbol: SLC25A22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191060.1 | NP_001177989.1 | 10 | 2362 | ||
| NM_001191061.1 | NP_001177990.1 | 10 | 2031 | |||
| NM_024698.5 | NP_078974.1 | 10 | 2199 | |||
| XM_011520369.1 | XP_011518671.1 | 10 | 2073 | |||
| XM_011520370.1 | XP_011518672.1 | 10 | 2141 | |||
| XM_011520371.1 | XP_011518673.1 | 10 | 2026 | |||
| GenBank mRNA | AJ428202.1 | - | 9 | 1919 | ||
| AK023106.1 | - | 9 | 1919 | |||
| AL832423.1 | - | 5 | 1451 | |||
| BC019033.2 | - | 10 | 1997 | 4577143 | ||
| BC023545.2 | - | 10 | 2157 | 4110556 | ||
| BC024212.2 | - | 6 | 1540 | |||

SLC25A22
Genomic Map