siRNA Details
| Gene Symbol: ASB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024701.3 | NP_078977.2 | 4 | 447 | ||
| NR_024581.1 | 3 | 296 | ||||
| NR_037164.1 | 4 | 447 | ||||
| GenBank mRNA | AF403031.1 | - | 4 | 429 | ||
| AK023196.1 | - | 4 | 430 | |||
| AK091935.1 | - | 3 | 296 | |||
| AK292071.1 | - | 4 | 433 | |||
| BC012056.1 | - | 4 | 424 | 4552643 | ||
| BG436652.1 | - | 3 | 273 | |||
| BP249999.1 | - | 4 | 447 | |||
| CR457302.1 | - | 4 | 403 | |||

ASB13
Genomic Map