siRNA Details
| Gene Symbol: C1orf115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024709.4 | NP_078985.3 | 2 | 1127 | ||
| GenBank mRNA | AK024208.1 | - | 2 | 1048 | ||
| AK056749.1 | - | 3 | 993 | |||
| AK091937.1 | - | 2 | 1061 | |||
| AK125403.1 | - | 2 | 1568 | |||
| BC036067.1 | - | 2 | 1093 | 5274863 | ||

C1orf115
Genomic Map