siRNA Details
| Gene Symbol: ELMO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024712.3 | NP_078988.2 | 8, 9 | 954 | ||
| GenBank mRNA | AK023886.1 | - | 7, 8 | 718 | ||
| AK056923.1 | - | 7, 8 | 917 | |||
| AK300976.1 | - | 8, 9 | 968 | |||
| BC034410.1 | - | 5, 6 | 566 | |||

ELMO3
Genomic Map