siRNA Details
| Gene Symbol: ELMO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024712.3 | NP_078988.2 | 15 | 1685 | ||
| GenBank mRNA | AK023886.1 | - | 14 | 1449 | ||
| AK056923.1 | - | 14 | 1648 | |||
| AK300976.1 | - | 15 | 1699 | |||
| BC015524.1 | - | 5 | 495 | |||
| BC034410.1 | - | 12 | 1297 | |||

ELMO3
Genomic Map