siRNA Details
| Gene Symbol: SEPW1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003009.2 | NP_003000.1 | 6 | 694 | ||
| GenBank mRNA | AF015283.1 | - | 6 | 543 | ||
| AK026717.1 | - | 5 | 2256 | |||
| AK091604.1 | - | 1 | 2322 | |||
| BC000581.2 | - | 6 | 559 | 3162433 | ||
| BC032546.1 | - | 6 | 584 | 5535970 | ||
| BC039597.1 | - | 6 | 562 | 6015153 | ||
| BC047893.1 | - | 6 | 561 | 6204763 | ||
| U67171.1 | - | 6 | 554 | |||

SEPW1
Genomic Map