siRNA Details
| Gene Symbol: IQCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270585.1 | NP_001257514.1 | 7, 8 | 1075 | ||
| NM_024726.4 | NP_079002.3 | 7, 8 | 1287 | |||
| NR_073043.1 | 7, 8 | 1287 | ||||
| XM_011511864.1 | XP_011510166.1 | 7, 8 | 1073 | |||
| XM_011511865.2 | XP_011510167.1 | 7, 8 | 1310 | |||
| GenBank mRNA | AK054711.1 | - | 7, 8 | 1074 | ||
| AK295566.1 | - | 7, 8 | 1072 | |||
| BC028699.1 | - | 7, 8 | 1290 | 4839325 | ||
| BC043504.1 | - | 7, 8 | 1201 | 5193742 | ||

IQCA1
Genomic Map