siRNA Details
| Gene Symbol: RERGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286201.1 | NP_001273130.1 | 4 | 376 | ||
| NM_024730.3 | NP_079006.1 | 5 | 533 | |||
| NR_104413.1 | 4 | 273 | ||||
| GenBank mRNA | AA220227.1 | - | 4 | 220 | ||
| AK026308.1 | - | 5 | 459 | |||
| BC042888.1 | - | 5 | 520 | 5198817 | ||
| BX422875.2 | - | 4 | 324 | |||
| DT220095.1 | - | 5 | 533 | |||

RERGL
Genomic Map