siRNA Details
| Gene Symbol: KLHL36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303451.1 | NP_001290380.1 | 2, 3 | 211 | ||
| NM_024731.3 | NP_079007.2 | 2, 3 | 211 | |||
| XM_005256149.2 | XP_005256206.1 | 2, 3 | 425 | |||
| XM_017023674.1 | XP_016879163.1 | 2, 3 | 173 | |||
| XM_017023675.1 | XP_016879164.1 | 2, 3 | 193 | |||
| GenBank mRNA | AK022605.1 | - | 1, 2 | 64 | ||
| BC032400.1 | - | 2, 3 | 187 | 5210986 | ||
| BC033821.1 | - | 2, 3 | 197 | 5219021 | ||
| DA049618.1 | - | 2, 3 | 213 | |||

KLHL36
Genomic Map