siRNA Details
Gene Symbol: ZNF408 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184751.1 | NP_001171680.1 | 4, 5 | 651 | ||
NM_024741.2 | NP_079017.1 | 4, 5 | 936 | |||
GenBank mRNA | AF346626.1 | - | 4, 5 | 886 | ||
AK022889.1 | - | 4, 5 | 885 | |||
AK302180.1 | - | 4, 5 | 651 | |||
BC013355.2 | - | 4, 5 | 658 | 4040144 | ||
BC015708.2 | - | 4, 5 | 665 | 3911552 |
