siRNA Details
| Gene Symbol: ARMC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105247.1 | NP_001098717.1 | 1 | 945 | ||
| NM_001288767.1 | NP_001275696.1 | 3 | 825 | |||
| NM_001301820.1 | NP_001288749.1 | 2 | 960 | |||
| NM_024742.2 | NP_079018.1 | 1 | 945 | |||
| XM_006721091.2 | XP_006721154.1 | 2 | 1078 | |||
| GenBank mRNA | AK024430.1 | - | 1 | 438 | ||
| AK294897.1 | - | 2 | 867 | |||
| AK295790.1 | - | 3 | 825 | |||
| AY217348.1 | - | 1 | 945 | |||
| BC014945.1 | - | 2 | 350 | 4860377 | ||

ARMC5
Genomic Map