siRNA Details
| Gene Symbol: SHCBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324318.1 | NP_001311247.1 | 6 | 662 | ||
| NM_024745.4 | NP_079021.3 | 6 | 773 | |||
| NR_136738.1 | 6 | 776 | ||||
| XM_011523335.2 | XP_011521637.1 | 6 | 4307 | |||
| GenBank mRNA | AK025662.1 | - | 2 | 79 | ||
| AK055931.1 | - | 6 | 744 | |||
| AK225315.1 | - | 2 | 79 | |||
| AK292164.1 | - | 6 | 740 | |||
| AK310430.1 | - | 6 | 742 | |||
| AK315706.1 | - | 6 | 742 | |||
| BC030699.1 | - | 6 | 748 | 4829528 | ||

SHCBP1
Genomic Map