siRNA Details
| Gene Symbol: HPS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024747.5 | NP_079023.2 | 1 | 1150 | ||
| GenBank mRNA | AF536238.1 | - | 1 | 1115 | ||
| BC011594.1 | - | 1 | 1115 | 4578480 | ||
| BC014993.1 | - | 1 | 1127 | 4869397 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HPS6| Gene Symbol: HPS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024747.5 | NP_079023.2 | 1 | 1150 | ||
| GenBank mRNA | AF536238.1 | - | 1 | 1115 | ||
| BC011594.1 | - | 1 | 1115 | 4578480 | ||
| BC014993.1 | - | 1 | 1127 | 4869397 | ||