siRNA Details
| Gene Symbol: HPS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024747.5 | NP_079023.2 | 1 | 1631 | ||
| GenBank mRNA | AF536238.1 | - | 1 | 1596 | ||
| AK026154.1 | - | 1 | 424 | |||
| BC011594.1 | - | 1 | 1596 | 4578480 | ||
| BC014993.1 | - | 1 | 1608 | 4869397 | ||

HPS6
Genomic Map