siRNA Details
| Gene Symbol: GSTCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031720.3 | NP_001026890.2 | 3 | 589 | ||
| NM_024751.3 | NP_079027.2 | 3 | 510 | |||
| XM_005263222.3 | XP_005263279.1 | 3 | 777 | |||
| XM_011532248.2 | XP_011530550.1 | 3 | 733 | |||
| XM_011532249.2 | XP_011530551.1 | 3 | 595 | |||
| XM_011532252.2 | XP_011530554.1 | 3 | 967 | |||
| GenBank mRNA | AK023335.1 | - | 3 | 768 | ||
| AK225563.1 | - | 3 | 768 | |||
| AK292355.1 | - | 3 | 592 | |||
| BC032942.2 | - | 3 | 589 | 4838672 | ||
| BX648355.1 | - | 3 | 511 | |||

GSTCD
Genomic Map