siRNA Details
| Gene Symbol: TTC21B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024753.4 | NP_079029.3 | 4 | 434 | ||
| XM_006712761.1 | XP_006712824.1 | 4 | 377 | |||
| XM_011511872.2 | XP_011510174.1 | 4 | 377 | |||
| XM_017004967.1 | XP_016860456.1 | 4 | 377 | |||
| XM_017004969.1 | XP_016860458.1 | 2 | 77 | |||
| GenBank mRNA | AB073395.1 | - | 4 | 341 | ||
| AK057268.1 | - | 4 | 372 | |||
| AK292942.1 | - | 4 | 377 | |||
| BC055424.1 | - | 4 | 434 | 4837489 | ||
| DA798655.1 | - | 2 | 161 | |||

TTC21B
Genomic Map