siRNA Details
| Gene Symbol: PTCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284403.1 | NP_001271332.1 | 6 | 541 | ||
| NM_001284404.1 | NP_001271333.1 | 7 | 671 | |||
| NM_001284405.1 | NP_001271334.1 | 8 | 789 | |||
| NM_024754.4 | NP_079030.3 | 9 | 868 | |||
| XM_005248603.2 | XP_005248660.1 | 8 | 724 | |||
| XM_017009866.1 | XP_016865355.1 | 8 | 729 | |||
| XM_017009867.1 | XP_016865356.1 | 7 | 641 | |||
| GenBank mRNA | AK022660.1 | - | 7 | 630 | ||
| AK056761.1 | - | 9 | 1532 | |||
| AK298750.1 | - | 6 | 532 | |||
| AK303639.1 | - | 7 | 640 | |||
| AK303776.1 | - | 8 | 788 | |||
| BC018720.1 | - | 9 | 844 | 4552964 | ||
| CR457290.1 | - | 5 | 396 | |||

PTCD2
Genomic Map