siRNA Details
| Gene Symbol: PTCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024754.4 | NP_079030.3 | 5 | 514 | ||
| XM_005248601.2 | XP_005248658.1 | 5 | 508 | |||
| XM_005248603.2 | XP_005248660.1 | 4 | 370 | |||
| XM_017009866.1 | XP_016865355.1 | 4 | 375 | |||
| XM_017009868.1 | XP_016865357.1 | 4 | 370 | |||
| GenBank mRNA | AK022660.1 | - | 3 | 276 | ||
| AK056761.1 | - | 5 | 1178 | |||
| BC018720.1 | - | 5 | 490 | 4552964 | ||
| CR457290.1 | - | 1 | 42 | |||
| DA082506.1 | - | 5 | 508 | |||

PTCD2
Genomic Map