siRNA Details
| Gene Symbol: CSNK2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001895.3 | NP_001886.1 | 10 | 1080 | ||
| NM_177559.2 | NP_808227.1 | 11 | 1197 | |||
| NM_177560.2 | NP_808228.1 | 9 | 870 | |||
| GenBank mRNA | AB451279.1 | - | 9 | 804 | ||
| AB451411.1 | - | 9 | 804 | |||
| AK302583.1 | - | 9 | 759 | |||
| AK312535.1 | - | 11 | 1088 | |||
| BC011668.2 | - | 10 | 952 | 4121882 | ||
| BC050036.1 | - | 10 | 971 | 5298655 | ||
| BC053532.1 | - | 9 | 899 | 3908058 | ||
| BC071167.1 | - | 9 | 811 | 5772615 | ||
| BT019792.1 | - | 9 | 804 | |||
| DQ354583.1 | - | 10 | 946 | |||
| J02853.1 | - | 10 | 957 | |||
| M55265.1 | - | 10 | 952 | |||
| S53149.1 | - | 10 | 957 | |||
| Gene Symbol: CSNK2A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256686.1 | NP_001243615.1 | 1 | 1042 | ||

CSNK2A1
Genomic Map