siRNA Details
| Gene Symbol: EFCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024768.2 | NP_079044.2 | 8 | 2448 | ||
| XM_005247781.3 | XP_005247838.1 | 8 | 2476 | |||
| XM_011513161.2 | XP_011511463.1 | 8 | 1908 | |||
| XM_011513164.2 | XP_011511466.1 | 7 | 2521 | |||
| GenBank mRNA | AK022119.1 | - | 6 | 1722 | ||

EFCC1
Genomic Map