siRNA Details
| Gene Symbol: NHEJ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024782.2 | NP_079058.1 | 5 | 725 | ||
| GenBank mRNA | AJ972687.1 | - | 5 | 669 | ||
| AK022672.1 | - | 5 | 661 | |||
| BC008210.2 | - | 1 | 51 | |||
| BC012732.2 | - | 5 | 646 | 4026495 | ||
| BC030986.1 | - | 5 | 678 | 4076851 | ||
| CR457291.1 | - | 4 | 579 | |||

NHEJ1
Genomic Map