siRNA Details
| Gene Symbol: RNF122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024787.3 | NP_079063.2 | 6 | 818 | ||
| XM_017013846.1 | XP_016869335.1 | 6 | 714 | |||
| GenBank mRNA | AK022588.1 | - | 6 | 815 | ||
| BC093884.1 | - | 6 | 436 | 7939729 | ||
| BC101573.1 | - | 6 | 436 | 8069079 | ||

RNF122
Genomic Map