siRNA Details
| Gene Symbol: TM4SF20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024795.3 | NP_079071.2 | 2 | 269 | ||
| XM_011511876.2 | XP_011510178.1 | 2 | 175 | |||
| GenBank mRNA | AK026453.1 | - | 2 | 268 | ||
| AY358671.1 | - | 2 | 261 | |||
| BC035754.1 | - | 2 | 256 | 5760060 | ||
| BC137256.1 | - | 2 | 255 | 9021253 | ||
| BC137257.1 | - | 2 | 255 | 9021254 | ||
| DA926038.1 | - | 2 | 269 | |||

TM4SF20
Genomic Map