siRNA Details
| Gene Symbol: TUBAL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171864.1 | NP_001165335.1 | 4 | 728 | ||
| NM_024803.2 | NP_079079.1 | 4 | 848 | |||
| GenBank mRNA | AK025318.1 | - | 4 | 850 | ||
| AK296616.1 | - | 4 | 726 | |||
| BC098247.1 | - | 4 | 721 | 40005930 | ||
| BC099716.1 | - | 4 | 841 | 40005934 | ||
| BC105634.1 | - | 4 | 841 | 40005931 | ||

TUBAL3
Genomic Map