siRNA Details
| Gene Symbol: RBFA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024805.2 | NP_079081.2 | 5 | 691 | ||
| GenBank mRNA | AF261073.2 | - | 5 | 571 | ||
| AK024825.1 | - | 5 | 679 | |||
| BC014195.2 | - | 5 | 553 | 3938406 | ||
| BC057772.1 | - | 5 | 559 | 30337652 | ||

RBFA
Genomic Map