siRNA Details
| Gene Symbol: CPSF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136040.2 | NP_001129512.1 | 3 | 345 | ||
| NM_001142565.1 | NP_001136037.1 | 3 | 345 | |||
| NM_024811.3 | NP_079087.3 | 3 | 421 | |||
| XM_005274298.4 | XP_005274355.1 | 3 | 431 | |||
| XM_005274299.4 | XP_005274356.1 | 3 | 348 | |||
| XM_005274303.4 | XP_005274360.1 | 3 | 302 | |||
| XM_011545257.2 | XP_011543559.1 | 3 | 431 | |||
| XM_011545258.2 | XP_011543560.1 | 3 | 302 | |||
| XM_011545259.2 | XP_011543561.1 | 3 | 302 | |||
| XM_011545260.2 | XP_011543562.1 | 3 | 345 | |||
| XM_011545261.2 | XP_011543563.1 | 3 | 346 | |||
| XM_011545262.2 | XP_011543564.1 | 3 | 303 | |||
| XM_011545263.2 | XP_011543565.1 | 3 | 302 | |||
| XM_017018345.1 | XP_016873834.1 | 3 | 431 | |||
| GenBank mRNA | AJ275970.1 | - | 2 | 211 | ||
| AK092037.1 | - | 2 | 275 | |||
| AK096343.1 | - | 3 | 301 | |||
| AK294578.1 | - | 3 | 301 | |||
| AK307190.1 | - | 3 | 467 | |||
| AL832546.1 | - | 1 | 16 | |||
| AL832667.1 | - | 3 | 336 | |||
| AL833332.1 | - | 3 | 301 | |||
| BC018135.1 | - | 3 | 311 | 3913173 | ||
| BX537888.1 | - | 3 | 326 | |||
| DA194038.1 | - | 3 | 421 | |||
| DA804579.1 | - | 3 | 345 | |||

CPSF7
Genomic Map