siRNA Details
| Gene Symbol: MAP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039580.1 | NP_001034669.1 | 9 | 1517 | ||
| XM_006714306.3 | XP_006714369.1 | 9 | 1662 | |||
| XM_011532253.1 | XP_011530555.1 | 9 | 1720 | |||
| XM_011532254.2 | XP_011530556.1 | 9 | 1629 | |||
| XM_011532255.2 | XP_011530557.1 | 8 | 1368 | |||
| XM_017008616.1 | XP_016864105.1 | 9 | 1717 | |||
| XM_017008617.1 | XP_016864106.1 | 9 | 1534 | |||
| XM_017008618.1 | XP_016864107.1 | 9 | 1495 | |||
| GenBank mRNA | AK024730.1 | - | 6 | 916 | ||
| AK301075.1 | - | 9 | 1309 | |||
| AY690636.1 | - | 9 | 1517 | |||
| BC022852.1 | - | 9 | 1342 | |||
| BC128540.1 | - | 9 | 1429 | |||
| BC128541.1 | - | 9 | 1432 | |||
| BC146864.1 | - | 9 | 1429 | 9056792 | ||

MAP9
Genomic Map