siRNA Details
| Gene Symbol: CAAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167575.1 | NP_001161047.1 | 2 | 724 | ||
| NM_024828.3 | NP_079104.3 | 2 | 570 | |||
| GenBank mRNA | AK023719.1 | - | 2 | 544 | ||
| AK300739.1 | - | 2 | 338 | |||
| AK301673.1 | - | 2 | 268 | |||
| BC014658.1 | - | 2 | 546 | 4648932 | ||
| BC071953.1 | - | 2 | 469 | 5431506 | ||
| BP228742.1 | - | 2 | 570 | |||

CAAP1
Genomic Map