siRNA Details
| Gene Symbol: GGNBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024835.4 | NP_079111.1 | 9 | 1365 | ||
| XM_005257689.2 | XP_005257746.1 | 9 | 1347 | |||
| XM_011525265.2 | XP_011523567.1 | 8 | 1110 | |||
| XM_011525266.1 | XP_011523568.1 | 8 | 1233 | |||
| XM_017025107.1 | XP_016880596.1 | 8 | 1078 | |||
| GenBank mRNA | AF126964.1 | - | 8 | 1096 | ||
| AF268387.1 | - | 8 | 2343 | |||
| AK024883.1 | - | 9 | 1321 | |||
| AK124053.1 | - | 3 | 1736 | |||
| AK124084.1 | - | 3 | 1281 | |||
| AK290687.1 | - | 8 | 1121 | |||
| AK301614.1 | - | 6 | 653 | |||
| BC137491.1 | - | 9 | 1231 | 9021491 | ||
| CD557886.1 | - | 2 | 195 | |||

GGNBP2
Genomic Map