siRNA Details
| Gene Symbol: ZNF672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024836.2 | NP_079112.1 | 4 | 892 | ||
| XM_005270336.1 | XP_005270393.1 | 4 | 737 | |||
| GenBank mRNA | AK025954.1 | - | 4 | 739 | ||
| AK027679.1 | - | 2 | 286 | |||
| BC008872.1 | - | 4 | 547 | |||
| BC029310.1 | - | 4 | 790 | |||
| BC035140.1 | - | 1 | 1272 | 5264126 | ||
| BC050348.1 | - | 1 | 1268 | 5260682 | ||
| BC068506.1 | - | 4 | 590 | 4824147 | ||

ZNF672
Genomic Map