siRNA Details
| Gene Symbol: THNSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024838.4 | NP_079114.3 | 3 | 846 | ||
| XM_005252597.2 | XP_005252654.1 | 3 | 922 | |||
| XM_017016665.1 | XP_016872154.1 | 4 | 932 | |||
| GenBank mRNA | AK094362.1 | - | 1 | 65 | ||
| AK125249.1 | - | 3 | 774 | |||
| BC035132.2 | - | 3 | 767 | 5263853 | ||
| BX647989.1 | - | 3 | 759 | |||

THNSL1
Genomic Map