siRNA Details
| Gene Symbol: SETD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160305.2 | NP_001153777.1 | 8 | 1269 | ||
| NM_024860.3 | NP_079136.2 | 9 | 1197 | |||
| NR_134583.1 | 9 | 1228 | ||||
| GenBank mRNA | AK024801.1 | - | 9 | 1153 | ||
| AK127136.1 | - | 5 | 2674 | |||
| AK290495.1 | - | 8 | 1231 | |||
| BC022451.1 | - | 8 | 1231 | 4794299 | ||
| BC110433.1 | - | 9 | 1212 | |||

SETD6
Genomic Map