siRNA Details
| Gene Symbol: DOK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308235.1 | NP_001295164.1 | 6 | 1517 | ||
| NM_001308236.1 | NP_001295165.1 | 6 | 1340 | |||
| NM_024872.2 | NP_079148.2 | 6 | 1296 | |||
| XM_005265985.3 | XP_005266042.1 | 6 | 1353 | |||
| XM_011534654.2 | XP_011532956.1 | 6 | 1230 | |||
| XM_011534655.2 | XP_011532957.1 | 6 | 1406 | |||
| XM_011534656.2 | XP_011532958.1 | 6 | 1207 | |||
| XM_011534657.2 | XP_011532959.1 | 5 | 1057 | |||
| XM_017009883.1 | XP_016865372.1 | 4 | 1028 | |||
| GenBank mRNA | AK026223.1 | - | 6 | 1291 | ||

DOK3
Genomic Map