siRNA Details
| Gene Symbol: CNTD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024877.3 | NP_079153.2 | 5 | 1322 | ||
| GenBank mRNA | AK023327.1 | - | 4 | 1296 | ||
| BM709160.1 | - | 1 | 389 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CNTD2| Gene Symbol: CNTD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024877.3 | NP_079153.2 | 5 | 1322 | ||
| GenBank mRNA | AK023327.1 | - | 4 | 1296 | ||
| BM709160.1 | - | 1 | 389 | |||