siRNA Details
| Gene Symbol: SLC35E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024881.4 | NP_079157.3 | 2, 3 | 516 | ||
| GenBank mRNA | AK024313.1 | - | 2, 3 | 343 | ||
| AK027699.1 | - | 2, 3 | 310 | |||
| BC062562.1 | - | 2, 3 | 516 | 6598965 | ||
| BX640756.1 | - | 1, 2 | 31 | |||
| CR457326.1 | - | 1, 2 | 66 | |||

SLC35E1
Genomic Map