siRNA Details
| Gene Symbol: ERMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024896.2 | NP_079172.2 | 8 | 1448 | ||
| XM_005251587.1 | XP_005251644.1 | 8 | 1419 | |||
| XM_017015139.1 | XP_016870628.1 | 9 | 1241 | |||
| GenBank mRNA | AB058718.2 | - | 8 | 1325 | ||
| AK093217.1 | - | 7 | 815 | |||
| AK127218.1 | - | 8 | 1437 | |||
| AK172831.1 | - | 2 | 152 | |||
| AL834191.1 | - | 7 | 1080 | |||
| BC136771.1 | - | 8 | 1422 | 9020761 | ||
| BC136773.1 | - | 8 | 1422 | 9020763 | ||
| BC144404.1 | - | 8 | 1422 | |||

ERMP1
Genomic Map