siRNA Details
| Gene Symbol: CEP76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024899.3 | NP_079175.2 | 4 | 544 | ||
| NR_073537.1 | 4 | 539 | ||||
| XM_005258149.4 | XP_005258206.1 | 4 | 523 | |||
| XM_017025981.1 | XP_016881470.1 | 4 | 523 | |||
| XM_017025982.1 | XP_016881471.1 | 4 | 604 | |||
| XM_017025983.1 | XP_016881472.1 | 4 | 523 | |||
| XM_017025986.1 | XP_016881475.1 | 3 | 281 | |||
| GenBank mRNA | AK022604.1 | - | 4 | 406 | ||
| AK301964.1 | - | 3 | 281 | |||
| BC026307.1 | - | 4 | 471 | 4792211 | ||
| BC033026.1 | - | 4 | 406 | |||

CEP76
Genomic Map