siRNA Details
| Gene Symbol: DENND2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271833.1 | NP_001258762.1 | 4 | 552 | ||
| NM_024901.4 | NP_079177.2 | 4 | 620 | |||
| XM_006710921.2 | XP_006710984.2 | 5 | 574 | |||
| XM_006710922.1 | XP_006710985.1 | 4 | 564 | |||
| XM_006710923.1 | XP_006710986.1 | 4 | 552 | |||
| XM_006710924.1 | XP_006710987.1 | 4 | 329 | |||
| XM_011542187.1 | XP_011540489.1 | 5 | 585 | |||
| XM_011542189.1 | XP_011540491.1 | 4 | 579 | |||
| XM_011542190.2 | XP_011540492.1 | 5 | 453 | |||
| XM_017002386.1 | XP_016857875.1 | 4 | 329 | |||
| XM_017002387.1 | XP_016857876.1 | 4 | 345 | |||
| XM_017002388.1 | XP_016857877.1 | 4 | 579 | |||
| XM_017002389.1 | XP_016857878.1 | 5 | 574 | |||
| GenBank mRNA | AK026110.1 | - | 4 | 423 | ||
| AK301583.1 | - | 3 | 416 | |||
| AL713773.1 | - | 4 | 463 | |||
| BC004557.2 | - | 4 | 576 | 3957641 | ||
| BP270686.1 | - | 4 | 552 | |||

DENND2D
Genomic Map