siRNA Details
| Gene Symbol: WDR76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167941.1 | NP_001161413.1 | 2 | 255 | ||
| NM_024908.3 | NP_079184.2 | 2 | 365 | |||
| XM_005254680.3 | XP_005254737.1 | 2 | 328 | |||
| GenBank mRNA | AK023035.1 | - | 2 | 270 | ||
| AK290933.1 | - | 2 | 287 | |||
| BC025247.1 | - | 2 | 296 | 4560352 | ||
| BC051855.1 | - | 2 | 268 | 6141090 | ||
| BX374706.2 | - | 2 | 181 | |||
| CN259143.1 | - | 2 | 306 | |||
| DA439086.1 | - | 2 | 365 | |||
| EF011624.1 | - | 2 | 268 | |||

WDR76
Genomic Map