siRNA Details
| Gene Symbol: WDR76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167941.1 | NP_001161413.1 | 11 | 1446 | ||
| NM_024908.3 | NP_079184.2 | 11 | 1556 | |||
| XM_005254680.3 | XP_005254737.1 | 11 | 1519 | |||
| GenBank mRNA | AK023035.1 | - | 11 | 1461 | ||
| AK290933.1 | - | 11 | 1478 | |||
| BC025247.1 | - | 11 | 1487 | 4560352 | ||
| BC051855.1 | - | 11 | 1459 | 6141090 | ||
| EF011624.1 | - | 11 | 1459 | |||

WDR76
Genomic Map