siRNA Details
| Gene Symbol: ZNF767P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027788.1 | 4 | 875 | |||
| GenBank mRNA | AK022762.1 | - | 4 | 812 | ||
| AK295771.1 | - | 5 | 840 | |||
| AY359125.1 | - | 1 | 36 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF767P| Gene Symbol: ZNF767P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027788.1 | 4 | 875 | |||
| GenBank mRNA | AK022762.1 | - | 4 | 812 | ||
| AK295771.1 | - | 5 | 840 | |||
| AY359125.1 | - | 1 | 36 | |||