siRNA Details
| Gene Symbol: CSTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005213.3 | NP_005204.1 | 3 | 350 | ||
| GenBank mRNA | AK291308.1 | - | 3 | 266 | ||
| AV716538.1 | - | 3 | 364 | |||
| AV717631.1 | - | 3 | 260 | |||
| BC010379.1 | - | 3 | 270 | 4063525 | ||
| CR456914.1 | - | 3 | 221 | |||
| X05978.1 | - | 3 | 266 | |||

CSTA
Genomic Map