siRNA Details
| Gene Symbol: POF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307940.1 | NP_001294869.1 | 16 | 1834 | ||
| NM_024921.3 | NP_079197.3 | 16 | 1834 | |||
| XM_005262203.3 | XP_005262260.1 | 16 | 1747 | |||
| GenBank mRNA | AF309774.1 | - | 16 | 1834 | ||
| AK025039.1 | - | 12 | 1427 | |||
| AK025080.1 | - | 8 | 799 | |||
| AK026445.1 | - | 4 | 327 | |||
| AK128541.1 | - | 1 | 4025 | |||
| AK225193.1 | - | 8 | 799 | |||
| AK290360.1 | - | 16 | 1783 | |||
| AK314295.1 | - | 16 | 1753 | |||
| BC017500.1 | - | 16 | 1808 | 4905554 | ||

POF1B
Genomic Map