siRNA Details
| Gene Symbol: ANKRD53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115116.1 | NP_001108588.1 | 3, 4 | 811 | ||
| NM_024933.3 | NP_079209.3 | 3, 4 | 811 | |||
| XM_005264569.4 | XP_005264626.1 | 3, 4 | 1971 | |||
| GenBank mRNA | AK093479.1 | - | 3, 4 | 647 | ||
| BC035234.1 | - | 3, 4 | 811 | 5166484 | ||
| DB036445.1 | - | 2, 3 | 409 | |||

ANKRD53
Genomic Map