siRNA Details
| Gene Symbol: PCNX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014801.3 | NP_055616.3 | 31 | 5653 | ||
| XM_011544278.2 | XP_011542580.2 | 24 | 3264 | |||
| XM_017002391.1 | XP_016857880.1 | 30 | 5217 | |||
| XM_017002392.1 | XP_016857881.1 | 18 | 2503 | |||
| GenBank mRNA | AB007895.1 | - | 8 | 2569 | ||
| AK055374.1 | - | 12 | 1819 | |||
| AK074078.1 | - | 26 | 4725 | |||
| AK090784.1 | - | 7 | 1885 | |||
| AK097229.1 | - | 17 | 2228 | |||

PCNX2
Genomic Map