siRNA Details
| Gene Symbol: PCNX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014801.3 | NP_055616.3 | 26 | 4820 | ||
| XM_011544278.2 | XP_011542580.2 | 19 | 2431 | |||
| XM_017002391.1 | XP_016857880.1 | 25 | 4384 | |||
| XM_017002392.1 | XP_016857881.1 | 13 | 1670 | |||
| GenBank mRNA | AB007895.1 | - | 3 | 1736 | ||
| AK055374.1 | - | 7 | 986 | |||
| AK074078.1 | - | 21 | 3892 | |||
| AK090784.1 | - | 2 | 1052 | |||
| AK307812.1 | - | 13 | 1666 | |||

PCNX2
Genomic Map