siRNA Details
| Gene Symbol: PCNX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014801.3 | NP_055616.3 | 5 | 1789 | ||
| XM_006711816.3 | XP_006711879.1 | 5 | 1611 | |||
| XM_017002391.1 | XP_016857880.1 | 4 | 1353 | |||
| GenBank mRNA | AK074078.1 | - | 1 | 1040 | ||

PCNX2
Genomic Map